accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89694-45-1

IUPAC Name: (5-bromo-2-methoxyphenyl)boronic acid
Synonyms: 5-bromo-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 230.85
Molecular Formula: C7H8BBrO3
InChi Key: FVRLSHRAYPFXRQ-UHFFFAOYSA-N
SMILES: COC1=C(C=C(Br)C=C1)B(O)O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results