accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89694-46-2

IUPAC Name: (2-chloro-5-methoxyphenyl)boronic acid
Synonyms: 2-chloro-5-methoxyphenylboronic acid
Molecular Weight (g/mol): 186.4
Molecular Formula: C7H8BClO3
InChi Key: REFXAANPQCJZRY-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C(=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results