accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89719-91-5

IUPAC Name: 3,6-dichloro-2-(trifluoromethyl)pyridine
Synonyms: 3,6-dichloro-2-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 215.98
Molecular Formula: C6H2Cl2F3N
InChi Key: WDFIJOQQKFWJEA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(Cl)C=CC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results