accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89762-42-5

IUPAC Name: N,N-diethyl-4-[2-(pyridin-4-yl)diazen-1-yl]aniline
Synonyms: N,N-diethyl-4-[2-(pyridin-4-yl)diazen-1-yl]aniline
Molecular Weight (g/mol): 254.34
Molecular Formula: C15H18N4
InChi Key: FLPXBJDOCHBJCT-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)N=NC1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results