accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89785-84-2

IUPAC Name: sodium (2S,3S)-3-methyl-4,4,7-trioxo-3-[(1H-1,2,3-triazol-1-yl)methyl]-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: sodium (2S,3S)-3-methyl-4,4,7-trioxo-3-(1,2,3-triazol-1-ylmethyl)-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol): 322.27
Molecular Formula: C10H11N4NaO5S
InChi Key: RFMIKMMOLPNEDG-LEFOPYBQSA-M
SMILES: [Na+].C[C@]1(CN2C=CN=N2)[C@@H](N2C(CC2=O)S1(=O)=O)C([O-])=O
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results