accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3ONHCH3OCH3OH3COH3C

CAS RN 89805-39-0

IUPAC Name: bis[2-(3,4-dimethoxyphenyl)ethyl](methyl)azanium chloride
Synonyms: bis[2-(3,4-dimethoxyphenyl)ethyl](methyl)azanium chloride
Molecular Weight (g/mol): 395.92
Molecular Formula: C21H30ClNO4
InChi Key: GYBONEHHSUABAO-UHFFFAOYSA-N
SMILES: [Cl-].COC1=CC=C(CC[NH+](C)CCC2=CC=C(OC)C(OC)=C2)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results