CAS RN 89827-45-2
CAS RN 89827-45-2
IUPAC Name:
6-bromo-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms:
6-bromo-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol):
247.09
Molecular Formula:
C12H7BrO
InChi Key:
DYTYBRPMNQQFFL-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC2=C1OC1=C2C=CC=C1
1
–
1
of
1
results
1
4-Bromodibenzofuran 98.0+%, TCI America™

CAS: 89827-45-2 Molecular Formula: C12H7BrO Molecular Weight (g/mol): 247.091 MDL Number: MFCD00185685 InChI Key: DYTYBRPMNQQFFL-UHFFFAOYSA-N PubChem CID: 458256 IUPAC Name: 4-bromodibenzofuran SMILES: C1=CC=C2C(=C1)C3=C(O2)C(=CC=C3)Br