accessibility menu, dialog, popup

Test

Viewing profile as user:

CAS RN 898288-91-0

CAS RN 898288-91-0

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 4-(1,3-oxazol-5-yl)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 4-(1,3-oxazol-5-yl)benzoate
Molecular Weight (g/mol): 355.22
Molecular Formula: C16H6F5NO3
InChi Key: FWUPBFHGEDUEID-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC=C(C=C2)C2=CN=CO2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results