accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 898289-26-4

IUPAC Name: methyl 2-(thiomorpholin-4-yl)pyridine-4-carboxylate
Synonyms: methyl 2-(thiomorpholin-4-yl)pyridine-4-carboxylate
Molecular Weight (g/mol): 238.31
Molecular Formula: C11H14N2O2S
InChi Key: PKBDUKGWOJJNRV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=NC=C1)N1CCSCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results