accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOOH

CAS RN 898289-59-3

IUPAC Name: 3-(2-methyl-1H-imidazol-1-yl)benzoic acid
Synonyms: 3-(2-methylimidazol-1-yl)benzoic acid
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: ZGRKUYBDLZMGQH-UHFFFAOYSA-N
SMILES: CC1=NC=CN1C1=CC=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results