accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 898289-62-8

IUPAC Name: methyl 2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoate
Synonyms: methyl 2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]benzoate
Molecular Weight (g/mol): 284.24
Molecular Formula: C13H11F3N2O2
InChi Key: VBXXEOGWKRJBBH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1C1=CC(=NN1C)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results