accessibility menu, dialog, popup

UserName

Viewing profile as user:
BCH3CH3N

CAS RN 89878-14-8

IUPAC Name: 3-(diethylboranyl)pyridine
Synonyms: 3-(diethylboranyl)pyridine
Molecular Weight (g/mol): 147.03
Molecular Formula: C9H14BN
InChi Key: OJKBCQOJVMAHDX-UHFFFAOYSA-N
SMILES: CCB(CC)C1=CN=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)