accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOONNH2

CAS RN 89937-52-0

IUPAC Name: 4-(2-aminoethyl)-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-(2-aminoethyl)-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 178.25
Molecular Formula: C6H14N2O2S
InChi Key: NICIHZYGEQHDPN-UHFFFAOYSA-N
SMILES: NCCN1CCS(=O)(=O)CC1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results