CAS RN 89976-27-2
IUPAC Name:
methyl 4-iodo-3-nitrobenzoate
Synonyms:
methyl 4-iodo-3-nitrobenzoate
Molecular Weight (g/mol):
307.04
Molecular Formula:
C8H6INO4
InChi Key:
SCMBIQRYVKITCY-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(I)C(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
Methyl 4-iodo-3-nitrobenzoate, 99%
Methyl 4-iodo-3-nitrobenzoate, 99%
CAS: 89976-27-2 Molecular Formula: C8H6INO4 Molecular Weight (g/mol): 307.043 MDL Number: MFCD08064223 InChI Key: SCMBIQRYVKITCY-UHFFFAOYSA-N PubChem CID: 297884 IUPAC Name: methyl 4-iodo-3-nitrobenzoate SMILES: COC(=O)C1=CC(=C(C=C1)I)[N+](=O)[O-]