accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89981-75-9

IUPAC Name: (1S)-1-(2,4-dichlorophenyl)ethan-1-aminium
Synonyms: (1S)-1-(2,4-dichlorophenyl)ethanaminium
Molecular Weight (g/mol): 191.07
Molecular Formula: C8H10Cl2N
InChi Key: OUVZHZAOWDHBOU-YFKPBYRVSA-O
SMILES: C[C@H]([NH3+])C1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results