CAS RN 90-04-0
o-Anisidine, 99%
CAS: 90-04-0 Molecular Formula: C7H9NO Molecular Weight (g/mol): 123.155 MDL Number: MFCD00007688 InChI Key: VMPITZXILSNTON-UHFFFAOYSA-N Synonym: o-anisidine,2-anisidine,ortho-anisidine,2-aminoanisole,o-methoxyaniline,o-aminoanisole,benzenamine, 2-methoxy,2-methoxybenzenamine,o-methoxyphenylamine,o-anisylamine PubChem CID: 7000 ChEBI: CHEBI:82288 IUPAC Name: 2-methoxyaniline SMILES: COC1=CC=CC=C1N
o-Anisidine, 98+%
CAS: 90-04-0 Molecular Formula: C7H9NO Molecular Weight (g/mol): 123.15 MDL Number: MFCD00007688 InChI Key: VMPITZXILSNTON-UHFFFAOYSA-N Synonym: o-anisidine,2-anisidine,ortho-anisidine,2-aminoanisole,o-methoxyaniline,o-aminoanisole,benzenamine, 2-methoxy,2-methoxybenzenamine,o-methoxyphenylamine,o-anisylamine PubChem CID: 7000 ChEBI: CHEBI:82288 IUPAC Name: 2-methoxyaniline SMILES: COC1=CC=CC=C1N
o-Anisidine 98.0+%, TCI America™
CAS: 90-04-0 Molecular Formula: C7H9NO Molecular Weight (g/mol): 123.155 MDL Number: MFCD00007688 InChI Key: VMPITZXILSNTON-UHFFFAOYSA-N Synonym: o-anisidine,2-anisidine,ortho-anisidine,2-aminoanisole,o-methoxyaniline,o-aminoanisole,benzenamine, 2-methoxy,2-methoxybenzenamine,o-methoxyphenylamine,o-anisylamine PubChem CID: 7000 ChEBI: CHEBI:82288 IUPAC Name: 2-methoxyaniline SMILES: COC1=CC=CC=C1N
o-Anisidine, SPEX CertiPrep™
CAS: 90-04-0 Molecular Formula: C7H9NO Molecular Weight (g/mol): 123.155 InChI Key: VMPITZXILSNTON-UHFFFAOYSA-N PubChem CID: 7000 ChEBI: CHEBI:82288 IUPAC Name: 2-methoxyaniline SMILES: COC1=CC=CC=C1N