accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90-11-9

IUPAC Name: 1-bromonaphthalene
Synonyms: 1-bromonaphthalene
Molecular Weight (g/mol): 207.07
Molecular Formula: C10H7Br
InChi Key: DLKQHBOKULLWDQ-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (7)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (5)
  • (4)
  • (2)
  • (3)
Physical Form 
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (5)
  • (3)
  • (7)
  • (3)
  • (3)
  • (2)
  • (7)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (4)
  • (2)
  • (3)
  • (10)