accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClClCl

CAS RN 90-17-5

IUPAC Name: 2,2,2-trichloro-1-phenylethyl acetate
Synonyms: trichloromethyl phenyl carbinyl acetate
Molecular Weight (g/mol): 267.53
Molecular Formula: C10H9Cl3O2
InChi Key: JKRWZLOCPLZZEI-UHFFFAOYNA-N
SMILES: CC(=O)OC(C1=CC=CC=C1)C(Cl)(Cl)Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)