accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHO

CAS RN 90-46-0

IUPAC Name: 9H-xanthen-9-ol
Synonyms: xanthydrol
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: JFRMYMMIJXLMBB-UHFFFAOYSA-N
SMILES: OC1C2=CC=CC=C2OC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results