accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClO

CAS RN 90-60-8

IUPAC Name: 3,5-dichloro-2-hydroxybenzaldehyde
Synonyms: 3,5-dichloro-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 191.01
Molecular Formula: C7H4Cl2O2
InChi Key: FABVMBDCVAJXMB-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(Cl)C=C1C=O
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (4)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
  • (4)
  • (4)