accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 90-96-0

IUPAC Name: bis(4-methoxyphenyl)methanone
Synonyms: bis(4-methoxyphenyl)methanone
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: RFVHVYKVRGKLNK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)