accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 900-91-4

IUPAC Name: 3,3,3-triphenylpropanoic acid
Synonyms: 3,3,3-triphenylpropanoic acid
Molecular Weight (g/mol): 302.37
Molecular Formula: C21H18O2
InChi Key: XMSJLUKCGWQAHO-UHFFFAOYSA-N
SMILES: OC(=O)CC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)