accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNNNH2OPOHOOPOOOPOHOOHONaNa

CAS RN 9000-30-0

IUPAC Name: disodium hydrogen [({[5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
Synonyms: guar gum hydrolyzed guar cyamopsis tetragonoloba gum cyamopsis tetragonoloba symbiosome extract
Molecular Weight (g/mol): 535.15
Molecular Formula: C10H14N5Na2O12P3
InChi Key: JEKDCIBJADJZSK-UHFFFAOYNA-L
SMILES: [Na+].[Na+].NC1=C2N=CN(C3CC(O)C(COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C2=NC=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)