CAS RN 90001-07-3
IUPAC Name:
[(4-hydroxyphenyl)methyl]phosphonic acid
Synonyms:
(4-hydroxyphenyl)methylphosphonic acid
Molecular Weight (g/mol):
188.12
Molecular Formula:
C7H9O4P
InChi Key:
ZHBRSHSRMYZHLS-UHFFFAOYSA-N
SMILES:
OC1=CC=C(CP(O)(O)=O)C=C1
1
–
1
of
1
results
1
(4-Hydroxybenzyl)phosphonic Acid 98.0+%, TCI America™
CAS: 90001-07-3 Molecular Formula: C7H9O4P Molecular Weight (g/mol): 188.12 MDL Number: MFCD16877729 InChI Key: ZHBRSHSRMYZHLS-UHFFFAOYSA-N PubChem CID: 12029255 IUPAC Name: [(4-hydroxyphenyl)methyl]phosphonic acid SMILES: OC1=CC=C(CP(O)(O)=O)C=C1