accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOOS

CAS RN 90001-64-2

IUPAC Name: 1-benzothiophene-2-sulfonyl chloride
Synonyms: 1-benzothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 232.7
Molecular Formula: C8H5ClO2S2
InChi Key: FKIIVBOPAHICHQ-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results