accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 900015-48-7

IUPAC Name: 2-(pyrrolidin-1-yl)-1,3-thiazole-5-carbaldehyde
Synonyms: 2-(pyrrolidin-1-yl)-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 182.24
Molecular Formula: C8H10N2OS
InChi Key: CFFWKLLEEVWNLE-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(S1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results