accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90007-06-0

IUPAC Name: N'-hydroxy-2-phenoxyethanimidamide
Synonyms: N'-hydroxy-2-phenoxyethanimidamide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: BIIYCNCZOGOUJP-UHFFFAOYSA-N
SMILES: NC(COC1=CC=CC=C1)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results