CAS RN 9001-63-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lysozyme, MP Biomedicals™
Used for lysing E. coli and Streptomycetes for extraction purposes. Lysozyme, MP Biomedicals preferentially hydrolyzes the β-1,4 glucosidic linkages between N-acetylmuramic acid and N-acetylglucosamine. Enzyme activity: ≥20,000u/mg solid.