CAS RN 9001-67-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Neuraminidase, Clostridium perfringens
CAS: 9001-67-6 MDL Number: MFCD00131711 Synonym: Acyl-neuraminyl hydrolase,EC 3.2.1.18
2
Neuraminidase, Clostridium perfringens, 50% w/w sucrose, Thermo Scientific Chemicals
CAS: 9001-67-6 MDL Number: MFCD00131711 Synonym: Acyl-neuraminyl hydrolase,EC 3.2.1.18