CAS RN 9001-99-4
IUPAC Name:
Ribonuclease A
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
RNase A Solution, 10 mg/ml, Spectrum™ Chemical
CAS: 9001-99-4 MDL Number: MFCD00132181 IUPAC Name: Ribonuclease A