accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOH2NHClHCl

CAS RN 9002-29-3

IUPAC Name: 3-(4-benzyl-5-methyl-1,4-diazepane-1-carbonyl)bicyclo[3.3.1]nonan-9-amine dihydrochloride
Synonyms: 3-(4-benzyl-5-methyl-1,4-diazepane-1-carbonyl)bicyclo[3.3.1]nonan-9-amine dihydrochloride
Molecular Weight (g/mol): 442.47
Molecular Formula: C23H37Cl2N3O
InChi Key: DPEYHNFHDIXMNV-UHFFFAOYNA-N
SMILES: Cl.Cl.CC1CCN(CCN1CC1=CC=CC=C1)C(=O)C1CC2CCCC(C1)C2N
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)