CAS RN 9003-11-6
Poloxamer 188
Molecular Formula: C5H10O2 Synonym: Pluronic™ F-68; Polyoxyethylene-polyoxypropylene block copolymer
Poloxamer 182, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 124, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 407, NF, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 184, Surfactant, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 188, NF, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 181, Surfactant, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 188, NF, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 181, Surfactant, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 184, Surfactant, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 182, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Poloxamer 407, NF, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1
Medisca Poloxamer NF (407) (Pluronic F-127)
Poloxamer NF (407) (Pluronic F-127) comes as white prilled or cast solid. Odorless, or has a very mild odor.
Poloxamer 124, Spectrum™ Chemical
CAS: 9003-11-6 Molecular Formula: C5H10O2 Molecular Weight (g/mol): 102.13 InChI Key: RVGRUAULSDPKGF-UHFFFAOYNA-N IUPAC Name: 2-methylOxirane, Oxirane SMILES: C1CO1.CC1CO1