accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H2C

CAS RN 9003-27-4

IUPAC Name: 2-methylprop-1-ene
Synonyms: isobutylene 2-methylpropene (dimer)
Molecular Weight (g/mol): 56.11
Molecular Formula: C4H8
InChi Key: VQTUBCCKSQIDNK-UHFFFAOYSA-N
SMILES: CC(C)=C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results