accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 9004-35-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C6H7O5)nC2H3O
InChi Key: —
SMILES: —
Quantity 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)