accessibility menu, dialog, popup

UserName

Viewing profile as user:
(R)(S)(R)O(R)OO(S)ORgpO(R)O∗RgpRgp(S)O(R)(R)ORgpORgpO∗Rgpn

CAS RN 9004-36-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C12H14O10)n
InChi Key: —
SMILES: *-O[C@@H]1[C@@H](CO*)O[C@@H](O[C@@H]2[C@@H](CO*)OC(-*)[C@H](O*)[C@H]2O*)[C@H](O*)[C@H]1O*
Quantity 
  • (1)
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)