accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOCH3OOH3COOH3COOOOCH3OOCH3OOH3COOH3CH3COOOOCH3OOCH3OOCH3OOOOCH3OOCH3OOCH3OOCH3OHOOOHOOHOHOHOOHHOHO

CAS RN 9004-39-1

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results