accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 9005-66-7

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula: (C2H4O)w(C2H4O)x(C2H4O)z(C2H4O)yC22H42O6
InChi Key: DVRFSZFBSOKHQW-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (5)
  • (4)
  • (6)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)