accessibility menu, dialog, popup

UserName

Viewing profile as user:
OMeOOOOOHOOHOOOH3Cwyzx(Z)(Z)

CAS RN 9005-70-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
InChi Key: ZGEQUSWSYZZNAI-CLFAGFIQNA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)