CAS RN 90050-59-2
IUPAC Name:
5-bromo-2-methylbenzaldehyde
Synonyms:
5-bromo-2-methylbenzaldehyde
Molecular Weight (g/mol):
199.05
Molecular Formula:
C8H7BrO
InChi Key:
YILPAIKZHXATHY-UHFFFAOYSA-N
SMILES:
CC1=C(C=O)C=C(Br)C=C1
1
–
1
of
1
results
1
5-Bromo-2-methylbenzaldehyde 95.0+%, TCI America™
CAS: 90050-59-2 Molecular Formula: C8H7BrO Molecular Weight (g/mol): 199.047 MDL Number: MFCD05662390 InChI Key: YILPAIKZHXATHY-UHFFFAOYSA-N PubChem CID: 10420289 IUPAC Name: 5-bromo-2-methylbenzaldehyde SMILES: CC1=C(C=C(C=C1)Br)C=O