CAS RN 9006-26-2
IUPAC Name:
2,5-dihydrofuran-2,5-dione; ethene
Synonyms:
ethylene/MA copolymer
Molecular Weight (g/mol):
126.11
Molecular Formula:
C6H6O3
InChi Key:
YYXLGGIKSIZHSF-UHFFFAOYSA-N
SMILES:
C=C.O=C1OC(=O)C=C1
1
–
1
of
1
results
1
Sigma Aldrich Poly(ethylene-alt-maleic anhydride)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.