accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 90076-67-8

IUPAC Name: potassium trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: potassium bistriflylimide anion
Molecular Weight (g/mol): 319.23
Molecular Formula: C2F6KNO4S2
InChi Key: KVFIZLDWRFTUEM-UHFFFAOYSA-N
SMILES: [K+].FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)