accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 900806-58-8

IUPAC Name: 5,13-diphenyl-8,16-dithiatetracyclo[7.7.0.0²,⁷.0¹⁰,¹⁵]hexadeca-1(9),2,4,6,10(15),11,13-heptaene
Synonyms: 5,13-diphenyl-8,16-dithiatetracyclo[7.7.0.0²,⁷.0¹⁰,¹⁵]hexadeca-1(9),2,4,6,10(15),11,13-heptaene
Molecular Weight (g/mol): 392.53
Molecular Formula: C26H16S2
InChi Key: SBJIDUSVEICMRY-UHFFFAOYSA-N
SMILES: S1C2=CC(=CC=C2C2=C1C1=C(S2)C=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results