accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOONHNHOCl

CAS RN 90098-04-7

IUPAC Name: 2-[(4-chlorophenyl)formamido]-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid
Synonyms: rebamipide
Molecular Weight (g/mol): 370.79
Molecular Formula: C19H15ClN2O4
InChi Key: ALLWOAVDORUJLA-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC(=O)NC2=CC=CC=C12)NC(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results