CAS RN 90101-20-5
CAS RN 90101-20-5
IUPAC Name:
tert-butyl N-(4-methylpyridin-2-yl)carbamate
Synonyms:
tert-butyl N-(4-methylpyridin-2-yl)carbamate
Molecular Weight (g/mol):
208.26
Molecular Formula:
C11H16N2O2
InChi Key:
SABMAMDNSRZJBK-UHFFFAOYSA-N
SMILES:
CC1=CC(NC(=O)OC(C)(C)C)=NC=C1
1
–
1
of
1
results
1
Sigma Aldrich tert-Butyl (4-methylpyridin-2-yl)carbamate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.