CAS RN 9011-15-8
CAS RN 9011-15-8
IUPAC Name:
butyl 2-methylprop-2-enoate
Synonyms:
butyl methacrylate polyisobutyl methacrylate
Molecular Weight (g/mol):
142.2
Molecular Formula:
C8H14O2
InChi Key:
SOGAXMICEFXMKE-UHFFFAOYSA-N
SMILES:
CCCCOC(=O)C(C)=C
1
–
1
of
1
results
1
Isobutyl Methacrylate Polymer, TCI America™
CAS: 9011-15-8 Molecular Formula: C8H14O2 MDL Number: MFCD00008931 Synonym: Methacrylic Acid Isobutyl Ester Polymer