accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOHOHOOH

CAS RN 90111-58-3

IUPAC Name: 2-[4-(dihydroxyboranyl)phenyl]acetic acid
Synonyms: [4-(dihydroxyboranyl)phenyl]acetic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C8H9BO4
InChi Key: NFGJQVPDPIGBJE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(CC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results