CAS RN 9012-36-6
Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
CAS: 9012-36-6 Molecular Formula: C12H18O9
Agarose (Low-Melting, <1kb DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
Agarose, C12H18O9, CAS Number-9012-36-6, 100g, Gray, Cream, DNase free, 0.15 max., Pass Test, 306.12, 35 deg.C max., 500g/cm2min., Genetic Analysis, Low-Melting, <1kb DNA/RNA, Poly Bottle, Powder/Solid, DNase- and RNase-Free, RT, 0.15% max. | CAS: 9012-36-6 | C12H18O9
Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™
CAS: 9012-36-6 Molecular Formula: C12H18O9
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
CAS: 9012-36-6 Molecular Formula: C12H18O9
Agarose (Medium-EEO/Protein Electrophoresis Grade), Fisher BioReagents
CAS: 9012-36-6 Molecular Formula: C12H18O9
Thermo Scientific Chemicals Agarose, Electrophoresis Grade
CAS: 9012-36-6 MDL Number: MFCD00081294
Agarose LE, Molecular Biology Grade, Ultrapure
CAS: 9012-36-6 MDL Number: MFCD00081294
Agarose (Intermediate Melting/PCR Grade), Fisher BioReagents
CAS: 9012-36-6 Molecular Formula: C12H18O9
Thermo Scientific Chemicals Agarose, high melting temperature, medium resolution
CAS: 9012-36-6 MDL Number: MFCD00081294
Thermo Scientific Chemicals Agarose, low gelling temperature, Molecular Biology Reagent
CAS: 9012-36-6 MDL Number: MFCD00081294
Thermo Scientific Chemicals Agarose, high gelling temperature, Molecular Biology Reagent
CAS: 9012-36-6 MDL Number: MFCD00081294
Agarose, Electrophoresis Grade, MP Biomedicals™
CAS: 9012-36-6 MDL Number: MFCD00081294 Synonym: 3, 6-Anhydro-α-L-galacto-β-D-galactan,FastLane agarose
MP Biomedicals™ Basic Agarose Premier
Basic Agarose Premier, Analytical Grade
Thermo Scientific Chemicals Agarose, Genetic Technology Grade
CAS: 9012-36-6 MDL Number: MFCD00081294