accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOFFF

CAS RN 90176-80-0

IUPAC Name: 4-fluoro-2-(trifluoromethyl)benzaldehyde
Synonyms: 4-fluoro-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 192.11
Molecular Formula: C8H4F4O
InChi Key: NONOHEMDNFTKCZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C(=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results