accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFFFFFFFFFFFHO

CAS RN 90177-96-1

IUPAC Name: 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane-1,8-diol
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7-dodecafluorooctane-1,8-diol
Molecular Weight (g/mol): 362.12
Molecular Formula: C8H6F12O2
InChi Key: XZJPYETUABEQFI-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CO
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)