accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOOBr

CAS RN 90224-73-0

IUPAC Name: 5-bromo-2-methyl-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 5-bromo-2-methylisoindole-1,3-dione
Molecular Weight (g/mol): 240.06
Molecular Formula: C9H6BrNO2
InChi Key: BEIQHTQYTDPHLX-UHFFFAOYSA-N
SMILES: CN1C(=O)C2=C(C=C(Br)C=C2)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results